Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurexin 3/NRXN3 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299887PCP
Description
Neurexin 3/NRXN3 Polyclonal antibody specifically detects Neurexin 3/NRXN3 in Human samples. It is validated for ELISASpecifications
Neurexin 3/NRXN3 | |
Polyclonal | |
ELISA | |
C14orf60, chromosome 14 open reading frame 60, KIAA0743MGC176711, neurexin 3, neurexin III, Neurexin III-alpha, Neurexin III-beta, neurexin-3-beta | |
Produced in rabbits immunized with purified, recombinant Human NRXN3/Neurexin 3 (Accession#: NP_620426.2; Met 1-Thr 357). | |
0.1 mL | |
Cell Biology, Neuroscience | |
9369 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction