Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurexophilin-1 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298420IR
Description
Neurexophilin-1 Polyclonal specifically detects Neurexophilin-1 in Rat samples. It is validated for ELISA.Specifications
Neurexophilin-1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Rat | |
Purified |
ELISA | |
DyLight 755 | |
neurexophilin 1, neurexophilin-1, NPH1 | |
Produced in rabbits immunized with purified, recombinant Neurexophilin-1 (Q63366-1; Met1-Gly271). | |
0.1 mL | |
Neuroscience | |
30010 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction