Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurexophilin-1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298356MFV500
Description
Neurexophilin-1 Polyclonal antibody specifically detects Neurexophilin-1 in Mouse samples. It is validated for ELISASpecifications
Neurexophilin-1 | |
Polyclonal | |
ELISA | |
neurexophilin 1, neurexophilin-1, NPH1 | |
Produced in rabbits immunized with purified, recombinant Mouse Neurexophilin-1 (Uniprot#: Q61200; Ala22-Gly271) | |
0.1 mL | |
Neuroscience | |
30010 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction