Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuritin (CPG15), Mouse anti-Human, Rat, Clone: A6.1, Millipore Sigma™

Catalog No. MABN184025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN184025 25 μg
MABN1840100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN184025 Supplier MilliporeSigma Supplier No. MABN184025UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Neuritin (CPG15), clone A6.1, Cat. No. MABN1840, is a highly specific mouse monoclonal antibody that targets Neuritin and has been tested for use in Western Blotting.

Neuritin (UniProt: Q9NPD7) is encoded by the NRN1 (also known as NRN) gene (Gene ID: 51299) in human. Neuritin, a conserved, glycosylphosphatidylinositol (GPI)-anchored protein that plays a role in the development, survival and function of neurons. Its activity is induced by brain-derived neurotrophic factor (BDNF) and neurotrophin-3 (NT-3). It is highly expressed by induced and natural regulatory T cells (TREGs). It also promotes the maintenance of the TREGs pool by stabilizing Foxp3 expression and promoting the persistence of these cells. Deficiency of Neuritin is shown to result in an imbalance between central and peripheral TREG populations and functions. Neuritin has a wide variety of effects in the nervous system, such as promoting neurite outgrowth, modulating neurite outgrowth during neuronal differentiation, and protecting motor neuron axons. It can also promote dendritic growth, regulate synaptic plasticity, and stabilize active synapses, and promote synaptic maturation. Its expression in the hippocampus is shown to be induced by neuronal activity following chemical- or electrical-induced seizures, ischemia, and exercise. Chronic antidepressant treatment is reported to increase neuritin expression in rat brain. Neuritin is synthesized with a signal peptide (aa 1-27) and a propeptide (aa 28-116) that are cleaved off in the mature form (aa 117-142). (Ref.: Son, H., et al. (2012). Proc. Natl. Acad. Sci. USA. 109(28); 11378-83; Zhou, S., and Zhou, J (2014). Curr. Med. Chem. 21(10); 1212-19). Note: A6.1 hybridoma clone, and WT/KO validation, courtesy of Lauren Makuch and Dr. Elly Nedivi, Massachusetts Institute of Technology. Note: A6.1 hybridoma clone, and WT/KO validation, courtesy of Dr. Lauren Makuch and Dr. Elly Nedivi, Massachusetts Institute of Technology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuritin (CPG15)
Applications Western Blot
Classification Monoclonal
Clone A6.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias candidate plasticity gene 15;CPG15
Gene Symbols NRN1;NRN
Host Species Mouse
Immunogen GST-tagged full-length human recombinant Neuritin.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001265639
Test Specificity Clone A6.1 detects human and rat Neuritin.
Target Species Human, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.