Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuritin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984751
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4751 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4751 Supplier Bioss Supplier No. BS2464R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

As the nervous system of a complex organism develops, it establishes functional networks through the growth and retraction of synaptic connections from growing axons and dendrites. This synaptic remodeling involves neuro-transmitter signaling, activation of neurotrophin receptors and alterations in gene expression. One such gene whose expression is increased by neural activity is neuritin, a GPI-anchored protein that is expressed in postmitotic differentiating neurons of the developing nervous system. Its expression is also induced by the neurotrophins BDNF and NT-3. Purified recombinant neuritin promotes neurite outgrowth and arborization in primary embryonic neuronal cultures, suggesting that neuritin may play a role as a downstream effector of activity-induced neurite outgrowth. More recent experiments have shown that neuritin is required for the androgen-induced axonal elongation in motor neurons and is upregulated following spinal cord injury, suggesting that neuritin may also play a role in survival and axonal regeneration.

Specifications

Antigen Neuritin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NRN1
Gene Accession No. Q9NPD7
Gene Alias 0710008J23Rik; Candidate plasticity gene 15 protein; candidate plasticity-related gene 15; Cpg15; dJ380B8.2; Neuritin; neuritin 1; NRN; Nrn1
Gene Symbols NRN1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Nrn1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51299
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.