Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEURL Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299299MFV610
Description
NEURL Polyclonal antibody specifically detects NEURL in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
NEURL | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
h-neu, h-neuralized 1, neu, neu-1, neuralized (Drosophila)-like, neuralized homolog (Drosophila), neuralized-like protein 1A, NEURL1A, NEURL1neuralized-like (Drosophila), RING finger protein 67, RNF67bA416N2.1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NEURL. | |
0.1 mL | |
Neuroscience | |
9148 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction