Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NEURL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536495
Description
This antibody detects endogenous protein at a molecular weight of 62 kDa. Purity is >95% by SDS-PAGE.
NEURL is involved in the determination of cell fate in the neurogenic region of the embryo and plays a role in the determination of cell fate in the central nervous system. NEURL may act as a tumor suppressor whose inactivation could be associated with malignant progression of astrocytic tumors.
Specifications
NEURL1 | |
Polyclonal | |
Unconjugated | |
NEURL1 | |
bA416N2.1; E3 ubiquitin-protein ligase NEURL1; h neu; h neuralized 1; h-neu; h-neuralized 1; neu; neu-1; NEUR1; neuralized E3 ubiquitin protein ligase 1; neuralized homolog; neuralized homolog 1A; neuralized-like homolog; neuralized-like protein 1A; NEURL; Neurl1; NEURL1A; RING finger protein 67; RING-type E3 ubiquitin transferase NEURL1; RNF67 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
309459, 9148 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O76050 | |
NEURL1 | |
Synthetic peptide corresponding to amino acids 221-270 of human Neuralized-1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction