Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEURL1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558037
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558037 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558037 Supplier Invitrogen™ Supplier No. PA558037
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALIDVYGITD EVQLLESAFA DTLTPARLSQ ARFSACLPPS SHDAANFDNN ELENNQVVAK LGHL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen NEURL1B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEURL1B
Gene Accession No. A8MQ27
Gene Alias E3 ubiquitin-protein ligase NEURL1B; E3 ubiquitin-protein ligase NEURL3; hNeur2; neur2; neuralized 2 alternative protein; neuralized E3 ubiquitin protein ligase 1B; neuralized homolog 1B; neuralized-2; Neuralized-like protein 1B; neuralized-like protein 3; NEURL1B; NEURL3; RING-type E3 ubiquitin transferase NEURL1B
Gene Symbols NEURL1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NEURL1B. Recombinant protein control fragment (Product #RP-96992).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54492
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.