Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEURL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571797
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571797 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571797 Supplier Invitrogen™ Supplier No. PA571797
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NEURL3 Polyclonal Antibody for ELISA

E3 ubiquitin-protein ligase. Seems to utilize UBE2E1. In vitro, generates polyubiquitin chains via non-canonical lysine residues suggesting that it is not involved in tagging substrates for proteasomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEURL3
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene NEURL3
Gene Accession No. Q96EH8
Gene Alias 2010300P06Rik; E3 ubiquitin-protein ligase LINCR; E3 ubiquitin-protein ligase NEURL3; Lincr; lung inducible neuralized-related C3HC4 RING finger; lung-inducible neuralized-related C3CH4 RING domain protein; lung-inducible neuralized-related C3CH4 RING domain-containing protein; lung-inducible neuralized-related C3HC4 RING domain protein; neuralized E3 ubiquitin protein ligase 3; neuralized homolog 3 homolog (Drosophila); neuralized homolog 3 pseudogene; neuralized-like protein 3; Neurl3; RING-type E3 ubiquitin transferase NEURL3
Gene Symbols NEURL3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 234-262 amino acids from the C-terminal region of human NEURL3.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93082
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.