Learn More
Specifications
Specifications
| Preparation Method | Neuro2a Chloroquine treated/ untreated lysates are provided as positive and negative control for Western blot analysis in Autophagy research. The lysates are provided as one vial of treated and one vial of untreated samples. Use 10 μL / lane for a standard mini-gel blot (approx. 1 mg/mL). |
| Quantity | 2 Vials |
| Content And Storage | Aliquot and store at -20°C to +80°C. Avoid freeze/thaw cycles. |
| Product Line | Novus |
| Research Category | Autophagy, Cellular Markers, Chaperone Mediated Autophagy (CMA), mTOR Pathway, Signal Transduction, Ubiquitin Proteasome Pathway |
| Target | Neuro2a |
| Synonym | LC3 Lysate, lc3b autophagy marker, LC3B western blot |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
