Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuroblastoma Antibody (FMG25), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250528IR
Description
Neuroblastoma Monoclonal specifically detects Neuroblastoma in Human samples. It is validated for Flow Cytometry.Specifications
Neuroblastoma | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Human cutaneous T cell lymphoma (HUT 78), SPF female Balb/c mice were injected i.p with 2 x 10*5 cells in complete Freund's adjuvant. 30 days later a 2nd i.p injection of 2 x 10*5 cells without adjuvant was given. Fusion was 5 days | |
0.1 mL | |
Cancer | |
Human | |
Purified |
Flow Cytometry | |
FMG25 | |
Flow Cytometry | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction