Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurocan, N-terminal Rabbit anti-Mouse, Rat, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABT138425UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT1384-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT1384-25UL Supplier MilliporeSigma Supplier No. ABT138425UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Neurocan, N-terminal, Cat. No. ABT1384, is a rabbit polyclonal antibody that detects Neurocan core protein and has been tested for use in Immunofluorescence, Immunohistochemistry, and Radioimmunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurocan, N-terminal
Applications Immunofluorescence, Immunohistochemistry, Immunoradiometric Assay
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum with 0.05% sodium azide.
Gene Alias Neurocan core protein;245 kDa early postnatal core glycoprotein;Chondroitin sulfate proteoglycan 3
Gene Symbols Ncan;Cspg3
Host Species Rabbit
Immunogen GST-tagged recombinant fragement corresponding to 164 amino acids from the N-terminal region of rat Neurocan.
Molecular Weight of Antigen 135.55 kDa calculated.
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Neurocan in fetal and adult rat brain.
Target Species Mouse, Rat
Content And Storage Stable for 1 year at -20°C from date of receipt.
Product Type Antibodies
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.