Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neurocan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547848
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547848 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547848 Supplier Invitrogen™ Supplier No. PA547848
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, less than 10% cross-reactivity with recombinant mouse Neurocan is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

Neurocan is a gene encoding a large chondroitin sulfate proteoglycan, predominantly expressed in the central nervous system. Located on chromosome 19 in humans, neurocan is part of the lectican family of proteoglycans, integral to the extracellular matrix where they modulate neuronal cell adhesion and migration. This proteoglycan is essential in the formation of the neural network during development and in maintaining the plasticity of the extracellular matrix in adult brains. Neurocan undergoes proteolytic processing, resulting in various fragments that influence neuronal growth and synapse stabilization. Altered neurocan expression has been linked to several neurological conditions such as schizophrenia, bipolar disorder, and Alzheimer's disease, likely due to its role in synaptic function and integrity of the neural matrix. Understanding neurocan's exact roles in neurological health and disease could illuminate pathways for potential therapeutic interventions targeting matrix remodeling and neuronal connectivity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurocan
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Ncan
Gene Accession No. O14594
Gene Alias 245 kDa early postnatal core glycoprotein; C230035B04; Chondroitin sulfate proteoglycan 3; chondroitin sulfate proteoglycan 3 (neurocan); chondroitin sulfate proteoglycan 3, related sequence; Cspg3; Cspg3-rs; NCAN; NEUR; neurocan; neurocan core protein; neurocan proteoglycan; TGFbeta-induced transcript, brain; Tgfbit
Gene Symbols Ncan
Host Species Sheep
Immunogen CHO-derived recombinant human Neurocan Glu23-Cys1321.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1463
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.