Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurocan Recombinant Rabbit Monoclonal Antibody (JE54-15), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Thermo Scientific MA536178
Description
chondroitin sulfate proteoglycan that may play a role in cell adhesion and cell migration.
Specifications
Neurocan | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O14594 | |
Ncan | |
Recombinant protein within Human Neurocan aa 1-96 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
JE54-15 | |
Unconjugated | |
Ncan | |
245 kDa early postnatal core glycoprotein; C230035B04; Chondroitin sulfate proteoglycan 3; chondroitin sulfate proteoglycan 3 (neurocan); chondroitin sulfate proteoglycan 3, related sequence; Cspg3; Cspg3-rs; NCAN; NEUR; neurocan; neurocan core protein; neurocan proteoglycan; TGFbeta-induced transcript, brain; Tgfbit | |
Rabbit | |
Protein A | |
RUO | |
1463 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction