Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NeuroD1 Antibody (5D2), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | NeuroD1 |
---|---|
Clone | 5D2 |
Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Flow Cytometry, ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
NeuroD1 Monoclonal antibody specifically detects NeuroD1 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
NeuroD1 | |
Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
basic helix-loop-helix transcription factor, BETA2, BHF-1, BHLHA3, bHLHa3beta-cell E-box transactivator 2, Class A basic helix-loop-helix protein 3, MODY6, NeuroD, NeuroD1, neurogenic differentiation 1, neurogenic differentiation factor 1, neurogenic helix-loop-helix protein NEUROD, NIDDM | |
A synthesized peptide derived from Human NeuroD1 [UniProt Q13562] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
5D2 | |
Flow Cytometry, ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Neuroscience, Stem Cells | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
4760 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title