Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEUROD1 Mouse anti-Human, Mouse, Clone:6E9G12, Abnova™

Catalog No. 89334485 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-485 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-485 Supplier Abnova Corporation Supplier No. MAB17800
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human NEUROD1.

This gene encodes a member of the NeuroD family of basic helix-loop-helix (bHLH) transcription factors. The protein forms heterodimers with other bHLH proteins and activates transcription of genes that contain a specific DNA sequence known as the E-box. It regulates expression of the insulin gene, and mutations in this gene result in type II diabetes mellitus. [provided by RefSeq]

Specifications

Antigen NEUROD1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6E9G12
Conjugate Unconjugated
Description neurogenic differentiation 1
Dilution ELISA (1:10000)
Flow Cytometry
Immunocytochemistry
Immunohistochemistry
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene NEUROD1
Gene Alias BETA2, BHF-1, NEUROD, bHLHa3
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 26-91 of human NEUROD1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 4760
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.