Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NeuroD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585775
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585775 Supplier Invitrogen™ Supplier No. PA585775
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%). Positive Control: U87-MG, SK-N-SH, IMR32, SK-N-AS, mouse fetal brain, rat cerebellum. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

NeuroD1 is a transcriptional activator that acts as a differentiation factor during neurogenesis. It has been demonstrated to bind to the insulin gene E-box. Efficient DNA binding requires dimerization with another basic helix-loop-helix (bHLH) protein. Defects in NEUROD1 are a cause of maturity onset diabetes of the young type VI (MODY6). MODY6 is a form of non-insulin-dependent diabetes mellitus characterized by an autosomal dominant mode of inheritance, onset during young adulthood and a primary defect in insulin secretion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NeuroD1
Applications Western Blot
Classification Polyclonal
Concentration 1.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene NEUROD1
Gene Accession No. Q13562, Q60867, Q64289
Gene Alias Basic helix-loop-helix factor 1; basic helix-loop-helix transcription factor; Beta2; beta-cell E-box transactivator 2; beta-cell E-box transcriptional activator 2; BHF1; BHF-1; bHLHa3; Class A basic helix-loop-helix protein 3; MODY6; neuro d1; Neurod; Neurod1; neurogenic differentiation 1; neurogenic differentiation factor 1; neurogenic helix-loop-helix protein NEUROD; neuronal differentiation 1
Gene Symbols NEUROD1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of mouse NeuroD1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18012, 29458, 4760
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.