Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NeuroD2 Antibody (PCRP-NEUROD2-1G1), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314015APC
This item is not returnable.
View return policy
Description
NeuroD2 Monoclonal antibody specifically detects NeuroD2 in Human samples. It is validated for Flow CytometrySpecifications
| NeuroD2 | |
| Monoclonal | |
| APC | |
| BHLHA1, bHLHa1neurogenic differentiation factor 2, Class A basic helix-loop-helix protein 1, MGC26304, NDRFneurogenic basic-helix-loop-helix protein, NeuroD2, NeuroD-related factor, neurogenic differentiation 2 | |
| Recombinant full-length human NeuroD2 protein (Uniprot: Q15784) | |
| 0.1 mL | |
| Neuroscience | |
| 4761 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry | |
| PCRP-NEUROD2-1G1 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction