Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NeuroD2 Antibody (PCRP-NEUROD2-1G1), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314015MFV500
Description
NeuroD2 Monoclonal antibody specifically detects NeuroD2 in Human samples. It is validated for Flow CytometrySpecifications
NeuroD2 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
PCRP-NEUROD2-1G1 | |
Flow Cytometry | |
BHLHA1, bHLHa1neurogenic differentiation factor 2, Class A basic helix-loop-helix protein 1, MGC26304, NDRFneurogenic basic-helix-loop-helix protein, NeuroD2, NeuroD-related factor, neurogenic differentiation 2 | |
Recombinant full-length human NeuroD2 protein (Uniprot: Q15784) | |
0.1 mL | |
Neuroscience | |
4761 | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction