Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NeuroD2 Antibody (SR1838), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | NeuroD2 |
---|---|
Clone | SR1838 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
NeuroD2 Monoclonal antibody specifically detects NeuroD2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
NeuroD2 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
BHLHA1, bHLHa1neurogenic differentiation factor 2, Class A basic helix-loop-helix protein 1, MGC26304, NDRFneurogenic basic-helix-loop-helix protein, NeuroD2, NeuroD-related factor, neurogenic differentiation 2 | |
A synthesized peptide derived from human NeuroD2 (Uniprot #: Q15784) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
SR1838 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
4761 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title