Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NeuroD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540477
Description
Peptide sequence: AEVKEEGELG GEEEEEEEEE EGLDEAEGER PKKRGPKKRK MTKARLERSK Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
Members of the myogenic determination family are basic helix-loop-helix (bHLH) proteins that can be separated into two classes, both of which work together to activate DNA transcription. Class A proteins include the ubiquitously expressed E-box binding factors, namely E2A, ITF-2 and HEB, while class B proteins, such as MyoD, myogenin and Neuro D (BETA2), are transiently expressed and exhibit a much more limited tissue distribution. Neuro D2 (neurogenic differentiation 2), also known as NDRF, NEUROD2 or bHLHa1, is a 382 amino acid nuclear protein that contains one bHLH domain and functions to induce neurogenic differentiation, playing an important role in the maintenance and determination of cell fate.
Specifications
NeuroD2 | |
Polyclonal | |
Unconjugated | |
NEUROD2 | |
BHLHA1; Brain bHLH protein KW8; class A basic helix-loop-helix protein 1; NDF2; NDR2; NDRF; NeuroD2; neuroD-related factor; neurogenic basic-helix-loop-helix protein; neurogenic differentiation 2; neurogenic differentiation factor 2; neuronal differentiation 2 | |
Rabbit | |
Protein A | |
RUO | |
4761 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q15784 | |
NEUROD2 | |
Synthetic peptide directed towards the N-terminal of human NEUROD2 (aa 71-120). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction