Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NeuroD2 Recombinant Rabbit Monoclonal Antibody (JE50-57)
GREENER_CHOICE

Catalog No. PIMA536147
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536147 Supplier Invitrogen™ Supplier No. MA536147
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

NeuroD2 Recombinant Monoclonal Antibody for IHC (P), Western Blot

Members of the myogenic determination family are basic helix-loop-helix (bHLH) proteins that can be separated into two classes, both of which work together to activate DNA transcription. Class A proteins include the ubiquitously expressed E-box binding factors, namely E2A, ITF-2 and HEB, while class B proteins, such as MyoD, myogenin and Neuro D (BETA2), are transiently expressed and exhibit a much more limited tissue distribution. Neuro D2 (neurogenic differentiation 2), also known as NDRF, NEUROD2 or bHLHa1, is a 382 amino acid nuclear protein that contains one bHLH domain and functions to induce neurogenic differentiation, playing an important role in the maintenance and determination of cell fate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NeuroD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE50-57
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene NEUROD2
Gene Accession No. Q15784, Q62414, Q63689
Gene Alias BHLHA1; Brain bHLH protein KW8; class A basic helix-loop-helix protein 1; NDF2; NDR2; NDRF; NeuroD2; neuroD-related factor; neurogenic basic-helix-loop-helix protein; neurogenic differentiation 2; neurogenic differentiation factor 2; neuronal differentiation 2
Gene Symbols NEUROD2
Host Species Rabbit
Immunogen Recombinant protein within Human NeuroD2 aa 260-382.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18013, 4761, 54276
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.