Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEUROD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564964
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564964 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564964 Supplier Thermo Scientific Supplier No. PA564964
Add to Cart
Add to Cart

NEUROD4 Polyclonal Antibody

NEUROD4 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen NEUROD4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEUROD4
Gene Accession No. Q9HD90
Gene Alias AI846749; ath3; ATH-3; Atoh3; atonal homolog 3; atonal-like 3; atonal-like protein 3; bHLH domain; bHLHa4; Class A basic helix-loop-helix protein 4; helix-loop-helix protein mATH-3; mATH3; MATH-3; NeuroD4; neurod4 protein; neurogenic differentiation 4; neurogenic differentiation factor 4; neuronal basic helix-loop-helix transcription factor Ath3; neuronal differentiation 4; protein atonal homolog 3; zath3
Gene Symbols NEUROD4
Host Species Rabbit
Immunogen Recombinant Human NEUROD4. Recombinant protein control fragment (Product #RP-91625).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58158
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.