Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEUROD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569017
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569017 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569017 Supplier Invitrogen™ Supplier No. PA569017
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Rat: 100%.

Probably acts as a transcriptional activator. Mediates neuronal differentiation. Required for the regulation of amacrine cell fate specification in the retina.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEUROD4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NEUROD4
Gene Accession No. Q9HD90
Gene Alias AI846749; ath3; ATH-3; Atoh3; atonal homolog 3; atonal-like 3; atonal-like protein 3; bHLH domain; bHLHa4; Class A basic helix-loop-helix protein 4; helix-loop-helix protein mATH-3; mATH3; MATH-3; NeuroD4; neurod4 protein; neurogenic differentiation 4; neurogenic differentiation factor 4; neuronal basic helix-loop-helix transcription factor Ath3; neuronal differentiation 4; protein atonal homolog 3; zath3
Gene Symbols NEUROD4
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human NEUROD4.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58158
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.