Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NeuroD6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567782
Description
Predicted to react with Mouse and Rat samples.
NEUROD6 contains 1 basic helix-loop-helix (bHLH) domain. It activates E box-dependent transcription in collaboration with TCF3/E47 and may be a trans-acting factor involved in the development and maintenance of the mammalian nervous system. It transactivates the promoter of its own gene.
Specifications
NeuroD6 | |
Polyclonal | |
Unconjugated | |
NEUROD6 | |
ATH2; Atoh2; atonal homolog 2; bHLHa2; brain my051 protein; class A basic helix-loop-helix protein 2; Helix-loop-helix protein mATH-2; MATH2; Math-2; My051; Neurod6; neurogenic differentiation 6; neurogenic differentiation factor 6; neuronal differentiation 6; Nex; Nex1; Nex1m; protein atonal homolog 2; Protein NEX-1; RGD1562793 | |
Rabbit | |
affinity chromatography | |
RUO | |
11922, 500137, 63974 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P48986, Q96NK8 | |
NEUROD6 | |
Synthesized peptide derived from human NeuroD6. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction