Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurodye RH-461, pure, Thermo Scientific™

Catalog No. AC332850050
Click to view available options
Quantity:
5 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108641-88-9
Molecular Formula C25H37Br2N3
Molecular Weight (g/mol) 539.4
MDL Number MFCD11656011
InChI Key COTJMUJHRGTONC-UHFFFAOYSA-L
Synonym N-(4-Trimethylammoniumpropyl)-4-(4-(4-(diethylamino)phenyl)butadienyl)pyridinium diacetate, RH-46
PubChem CID 131865623
IUPAC Name 3-[4-[4-[4-(diethylamino)phenyl]buta-1,3-dienyl]pyridin-1-ium-1-yl]propyl-trimethylazanium;dibromide
SMILES CCN(CC)C1=CC=C(C=C1)C=CC=CC2=CC=[N+](C=C2)CCC[N+](C)(C)C.[Br-].[Br-]

Specifications

Formula Weight 497.67
Grade Extra Pure
Quantity 5 mg
Chemical Name or Material Neurodye RH-461
Hazard Category LIGHT SENSITIVE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.