Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neurofascin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547468
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547468 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547468 Supplier Invitrogen™ Supplier No. PA547468
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

This antibody detects human, mouse, rat Neurofascin in Western blots and rat Neurofascin in direct ELISAs. Reconstitute at 0.2 mg/mL in sterile PBS.

Neurofascin encodes an L1 family immunoglobulin cell adhesion molecule with multiple IGcam and fibronectin domains. The protein functions in neurite outgrowth, neurite fasciculation, and organization of the axon initial segment (AIS) and nodes of Ranvier on axons during early development. Both the AIS and nodes of Ranvier contain high densities of voltage-gated Na+ (Nav) channels which are clustered by interactions with cytoskeletal and scaffolding proteins including this protein, gliomedin, ankyrin 3 (ankyrin-G), and betaIV spectrin. This protein links the AIS extracellular matrix to the intracellular cytoskeleton. Neurofascin undergoes extensive alternative splicing, and the full-length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurofascin
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene NFASC
Gene Accession No. O94856, P97685, Q810U3
Gene Alias AA387016; D430023G06Rik; DKFZp686P2250; KIAA0756; KIAA0756FLJ46866; mKIAA0756; neurofascin; neurofascin homolog; NF; Nfasc; NRCAML
Gene Symbols NFASC
Host Species Chicken
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Neurofascin Ile25-Ala1031.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116690, 23114, 269116
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.