Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurofibromin 1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100418AF750
Description
Neurofibromin 1 Polyclonal antibody specifically detects Neurofibromin 1 in Human, Mouse, C. elegans samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Neurofibromin 1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS | |
The immunogen recognized by this antibody maps to a region between residue 2760 and the C-terminus (residue 2818) of human Neurofibromin 1 using the numbering given in entry NP_000258.1 (GeneID 4763) | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
4763 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, C. elegans | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction