Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurofibromin 1 Antibody (CL11773), Novus Biologicals™

Mouse Monoclonal Antibody
$330.00 - $547.00
Specifications
Antigen | Neurofibromin 1 |
---|---|
Clone | CL11773 |
Dilution | Immunocytochemistry/Immunofluorescence 2-10 ug/ml |
Applications | Immunofluorescence |
Classification | Monoclonal |
Description
Neurofibromin 1 Monoclonal antibody specifically detects Neurofibromin 1 in Human samples. It is validated for ImmunofluorescenceSpecifications
Neurofibromin 1 | |
Immunocytochemistry/Immunofluorescence 2-10 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS | |
This antibody was developed against a Synthetic Peptide of P21359. Actual immunogen sequence is proprietary. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL11773 | |
Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
PBS, pH 7.2, 40% glycerol | |
4763 | |
IgG2b | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title