Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofibromin 1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB243562 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB243562 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB243562 Supplier Novus Biologicals Supplier No. NB100418JF646
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Neurofibromin 1 Polyclonal antibody specifically detects Neurofibromin 1 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen Neurofibromin 1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2760 and the C-terminus (residue 2818) of human Neurofibromin 1 using the numbering given in entry NP_000258.1 (GeneID 4763)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4763
Target Species Human, Mouse, C. elegans
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.