Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofibromin 1 Antibody (McNFn27a), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB228884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB228884 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB228884 Supplier Novus Biologicals Supplier No. NBP280870AF350
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neurofibromin 1 Monoclonal antibody specifically detects Neurofibromin 1 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Neurofibromin 1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone McNFn27a
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS
Host Species Mouse
Immunogen A highly conserved N-terminal peptide, corresponding to amino acids 27-41 of human Neurofibromin 1. [UniProt# P21359]
Purification Method Protein A or G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4763
Target Species Human, Mouse, Rat, Guinea Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.