Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurofibromin 1 Antibody (McNFn27a), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB300153G
Description
Neurofibromin 1 Monoclonal specifically detects Neurofibromin 1 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Neurofibromin 1 | |
Monoclonal | |
DyLight 488 | |
P21359 | |
NF1 | |
A highly conserved N-terminal peptide, corresponding to amino acids 27-41 of human Neurofibromin 1. [UniProt# P21359] | |
0.2 mL | |
Cancer, Cell Cycle and Replication, Chromatin Research, Cytoskeleton Markers, Hypoxia, Neuroscience, Neurotransmission, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
4763 | |
Human, Mouse, Rat, Guinea Pig | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
McNFn27a | |
Western Blot, ELISA, Immunohistochemistry-Paraffin | |
DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
This is specific for full-length or truncated forms of neurofibromin. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction