Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 4763 |
| Protein Family | Neurofibromin 1 |
| Concentration | 1.0 mg/mL |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| For Use With (Application) | Antibody Competition |
| Gene Symbol | NF1 |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
| Research Category | Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Hypoxia, Neuroscience, Neurotransmission, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
