Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilament (NEFL) Monoclonal Antibody (OTI3C4), TrueMAB™, OriGene
SDP

Catalog No. 502543389
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3389 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3389 Supplier OriGene Supplier No. CF814272
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neurofilament (NEFL) Monoclonal antibody specifically detects Neurofilament (NEFL) in Human samples. It is validated for ELISA

Specifications

Antigen Neurofilament (NEFL)
Applications ELISA
Classification Monoclonal
Clone OTI3C4
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CR2
Gene Accession No. P20023
Gene Alias 68 kDa neurofilament protein; AI847934; CMT1F; CMT2E; FLJ53642; light molecular weight neurofilament protein; micro glutamic acid-rich protein; NEF3; Nefl; nefl.L; neurofilament; neurofilament 3; neurofilament L subunit; neurofilament light; neurofilament light polypeptide; neurofilament protein; neurofilament protein L; neurofilament protein, light chain; neurofilament subunit NF-L; neurofilament triplet L protein; neurofilament, light L homeolog; neurofilament, light polypeptide; neurofilament, light polypeptide 68kDa; neurofilament, light polypeptide L homeolog; neurofilament-l; neurofilament-light; Nf68; NFL; NF-L; NFM; NF-M; NLC; OTTHUMP00000196614; PPP1R110; protein phosphatase 1, regulatory subunit 110; XELAEV_18018673mg; XNF-L
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 208-587 of human CD21 (NP_001868) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1380
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.