Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilaments 70/200 kDa Monoclonal Antibody (2F11), Invitrogen™

Catalog No. PIMA515234
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515234 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515234 Supplier Thermo Scientific Supplier No. MA515234

Mouse Monoclonal Antibody

MA5-15234 targets Neurofilament (200kDa and 68kDa) in IF, IHC (P), IP, and WB applications and shows reactivity with Feline, Human, mouse, Rabbit, and Rat samples. The MA5-15234 immunogen is neurofilament purified from human brain.

Neurofilaments usually contain three intermediate filament proteins: NEFL, NEFM, and NEFH which are involved in the maintenance of neuronal caliber. NEFH has an important function in mature axons that is not subserved by the two smaller NF proteins. May additionally cooperate with the neuronal intermediate filament proteins PRPH and INA to form neuronal filamentous networks.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurofilaments 70/200 kDa
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2F11
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 15mM sodium azide
Gene NEFH
Gene Accession No. P07196, P08551, P12036, P16884, P19246, P19527
Gene Alias 200 kDa neurofilament protein; 68 kDa neurofilament protein; AI847934; CMT1F; CMT2CC; CMT2E; KIAA0845; light molecular weight neurofilament protein; LOW QUALITY PROTEIN: neurofilament light polypeptide; mKIAA0845; NEFH; Nefl; neurofilament 200kDa; neurofilament heavy; neurofilament heavy polypeptide; neurofilament light; neurofilament light polypeptide; neurofilament protein L; neurofilament protein, light chain; neurofilament subunit NF-L; neurofilament triplet H protein; Neurofilament triplet L protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, light polypeptide; neurofilament, light polypeptide 68kDa; neurofilament-H; NF200; NF68; NFH; NF-H; NFL; NF-L; PPP1R110; protein phosphatase 1, regulatory subunit 110
Gene Symbols NEFH, NEFL
Host Species Mouse
Immunogen Neurofilament purified from human brain
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100353082, 100357177, 101081039, 101087272, 18039, 24587, 380684, 4744, 4747, 83613
Target Species Feline, Human, Mouse, Rabbit, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.