Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NeuroG3 Monoclonal Antibody (1800CT113.128.46)
GREENER_CHOICE

Catalog No. PIMA537741
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537741 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537741 Supplier Invitrogen™ Supplier No. MA537741
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NeuroG3 Monoclonal Antibody for Western Blot

Neurog 3 protein contains one basic helix-loop-helix (bHLH) domain.It is both necessary and sufficient to induce endocrine islet cell differentiation from embryonic pancreatic progenitors. Not only does itinitiates endocrine cell differentiation, italso promotes islet cell maturation and maintains islet function. It is reported that Neurogenin 3 can redirect the differentiation of bipotential secretory progenitors to endocrine rather than goblet cell fate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NeuroG3
Applications Western Blot
Classification Monoclonal
Clone 1800CT113.128.46
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene NEUROG3
Gene Accession No. Q9Y4Z2
Gene Alias ATH4B; Atoh5; atonal homolog 5; BHLHA7; Class A basic helix-loop-helix protein 7; helix-loop-helix protein mATH-4B; LOW QUALITY PROTEIN: neurogenin-3; Math4B; Neurog3; neurogenin 3; Neurogenin 3 (Atonal protein homolog 5) (Helix-loop-helix protein mATH-4B) (MATH4B); neurogenin3; neurogenin-3; ngn3; NGN-3; Protein atonal homolog 5; Relax; transcriptional regulator, Relax
Gene Symbols NEUROG3
Host Species Mouse
Immunogen recombinant protein between 1-214 amino acids from human Neurogenin3.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50674
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.