Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEUROG3 Mouse anti-Human, Clone: OTI3B5, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500346S

Description
Neurog 3 protein contains one basic helix-loop-helix (bHLH) domain.It is both necessary and sufficient to induce endocrine islet cell differentiation from embryonic pancreatic progenitors. Not only does itinitiates endocrine cell differentiation, italso promotes islet cell maturation and maintains islet function. It is reported that Neurogenin 3 can redirect the differentiation of bipotential secretory progenitors to endocrine rather than goblet cell fate.Specifications
NEUROG3 | |
Monoclonal | |
1.3 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9Y4Z2 | |
NEUROG3 | |
Full-length protein expressed in 293T cell transfected with human NEUROG3 expression vector | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3B5 | |
Unconjugated | |
NEUROG3 | |
Atoh5, Math4B, NGN-3, bHLHa7, ngn3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
50674 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction