Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NeuroG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523481
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523481 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523481 Supplier Invitrogen™ Supplier No. PA523481
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NeuroG3 Polyclonal Antibody for IHC (P)

Neurog 3 protein contains one basic helix-loop-helix (bHLH) domain.It is both necessary and sufficient to induce endocrine islet cell differentiation from embryonic pancreatic progenitors. Not only does itinitiates endocrine cell differentiation, italso promotes islet cell maturation and maintains islet function. It is reported that Neurogenin 3 can redirect the differentiation of bipotential secretory progenitors to endocrine rather than goblet cell fate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NeuroG3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene NEUROG3
Gene Accession No. P70661
Gene Alias ATH4B; Atoh5; atonal homolog 5; BHLHA7; Class A basic helix-loop-helix protein 7; helix-loop-helix protein mATH-4B; LOW QUALITY PROTEIN: neurogenin-3; Math4B; Neurog3; neurogenin 3; Neurogenin 3 (Atonal protein homolog 5) (Helix-loop-helix protein mATH-4B) (MATH4B); neurogenin3; neurogenin-3; ngn3; NGN-3; Protein atonal homolog 5; Relax; transcriptional regulator, Relax
Gene Symbols NEUROG3
Host Species Rabbit
Immunogen A portion of amino acids 1-69 of mouse Neurog 3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11925
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.