Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurogenin-1 Antibody (OTI3F9), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272948AF594
Description
Neurogenin-1 Monoclonal specifically detects Neurogenin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Neurogenin-1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
NEUROG1 | |
Full length recombinant protein of human Neurogenin-1 (NP_006152) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3F9 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
AKA, BHLHA6, bHLHa6NGN-1, Class A basic helix-loop-helix protein 6, Math4C, NeuroD3, NEUROD3neurogenic differentiation 3, Neurogenic basic-helix-loop-helix protein, Neurogenic differentiation factor 3, neurogenin 1, neurogenin-1, NGN, NGN1 | |
Mouse | |
Affinity Purified | |
Neuroscience | |
4762.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction