Learn More
Invitrogen™ Neurogenin 1 Monoclonal Antibody (OTI9A8)

Description
Neurogenin 1 Monoclonal Antibody for ICC/IF, IHC (P), IP
Specifications
Specifications
| Antigen | Neurogenin 1 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI9A8 |
| Concentration | 0.75 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | NEUROG1 |
| Gene Accession No. | Q92886 |
| Gene Alias | AKA; ATH4C; basic helix-loop-helix transcription factor; bHLHa6; cb260; chunp6899; class A basic helix-loop-helix protein 6; Helix-loop-helix protein mATH-4C; Math4C; Neurod3; NEUROG1; Neurogenic basic-helix-loop-helix protein; neurogenic differentiation 3; neurogenic differentiation factor 3; neurogenin; neurogenin 1; neurogenin related protein-1; neurogenin-1; Neurogenin-related protein 1; NGN; Ngn1; NGN-1; ngr1; transcription factor; Zath4c; zNgn1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.