Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neurogenin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540478
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540478 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540478 Supplier Invitrogen™ Supplier No. PA540478
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ASDAESWGSG AAAASPLSDP SSPAASEDFT YRPGDPVFSF PSLPKDLLHT Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 79%; Rat: 93%.

Neurogenin 1 is a basic helix-loop-helix (bHLH) proteins are transcription factors involved in determining cell type during development. NeuroG1 is a bHLH protein with dual cell-fate specification roles. It functions during neurogenesis, and it has also been shown to inhibit the differentiation of neural stem cells into astrocytes. NeuroG1 promotes neurogenesis by functioning as a transcriptional activator, yet it inhibits astrocyte differentiation by compartmentalizing the CREB-binding protein transcription complex away from astrocyte differentiation genes and by inhibiting STAT transcription factors essential for gliogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurogenin 1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NEUROG1
Gene Accession No. P70660, Q92886
Gene Alias AKA; ATH4C; basic helix-loop-helix transcription factor; bHLHa6; cb260; chunp6899; class A basic helix-loop-helix protein 6; Helix-loop-helix protein mATH-4C; Math4C; Neurod3; NEUROG1; Neurogenic basic-helix-loop-helix protein; neurogenic differentiation 3; neurogenic differentiation factor 3; neurogenin; neurogenin 1; neurogenin related protein-1; neurogenin-1; Neurogenin-related protein 1; NGN; Ngn1; NGN-1; ngr1; transcription factor; Zath4c; zNgn1
Gene Symbols NEUROG1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human NEUROG1 (aa 180-229).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18014, 4762
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.