Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurogenin-2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Neurogenin-2 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Neurogenin-2 Polyclonal specifically detects Neurogenin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Neurogenin-2 | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
63973 | |
Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human NGN2. The immunogen is located within the last 50 amino acids of NGN2. Amino Acid Squence: CAGSDMDYWQPPPPDKHRYA | |
Primary | |
NGN2 antibody is predicted to not cross-react with neurogenin 1. |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
Atoh4, BHLHA8, bHLHa8MGC46562, Class A basic helix-loop-helix protein 8, Math4A, neurogenin 2, neurogenin-2, ngn-2, NGN2, Protein atonal homolog 4 | |
NEUROG2 | |
IgG | |
Affinity Purified | |
30 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title