Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurogenin-2 Antibody (SR1400), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322171100UL
Description
Neurogenin-2 Monoclonal antibody specifically detects Neurogenin-2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Neurogenin-2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1400 | |
Western Blot 1:500-1:2000 | |
Atoh4, BHLHA8, bHLHa8MGC46562, Class A basic helix-loop-helix protein 8, Math4A, neurogenin 2, neurogenin-2, ngn-2, NGN2, Protein atonal homolog 4 | |
A synthesized peptide derived from human Neurogenin-2 (Uniprot #: Q9H2A3) | |
100 μg | |
Neuroscience, Signal Transduction | |
63973 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction