Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurogenin-3 Antibody (PCRP-NEUROG3-1E10), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314029R
This item is not returnable.
View return policy
Description
Neurogenin-3 Monoclonal antibody specifically detects Neurogenin-3 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
| Neurogenin-3 | |
| Monoclonal | |
| DyLight 550 | |
| Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, neurogenin 3, neurogenin-3, NGN3, Protein atonal homolog 5 | |
| Recombinant human full-lengthNeurogenin-3protein (Uniprot: Q9Y4Z2) | |
| 0.1 mL | |
| Stem Cells | |
| 50674 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, ELISA, Immunofluorescence | |
| PCRP-NEUROG3-1E10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction