Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurogenin-3 Mouse anti-Human, Clone: PCRP-NEUROG3-1E10, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | Neurogenin-3 |
---|---|
Clone | PCRP-NEUROG3-1E10 |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array |
Applications | Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array |
Description
Neurogenin-3 Monoclonal antibody specifically detects Neurogenin-3 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
Neurogenin-3 | |
0.2 mg/ml | |
Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array | |
Unconjugated | |
Mouse | |
Stem Cells | |
10 mM PBS with 0.05% BSA | |
Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, neurogenin 3, neurogenin-3, NGN3, Protein atonal homolog 5 | |
Recombinant human full-lengthNeurogenin-3protein (Uniprot: Q9Y4Z2) | |
Primary | |
Store at 4°C. |
PCRP-NEUROG3-1E10 | |
Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q9Y4Z2 | |
50674 | |
IgG2b | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title