Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuroglycan C/CSPG5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Neuroglycan C/CSPG5 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Neuroglycan C/CSPG5 Polyclonal specifically detects Neuroglycan C/CSPG5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Neuroglycan C/CSPG5 | |
Polyclonal | |
Rabbit | |
Cellular Markers, Neuroscience | |
CALEB, chondroitin sulfate proteoglycan 5, chondroitin sulfate proteoglycan 5 (neuroglycan C), MGC44034, Neuroglycan C, NGCAcidic leucine-rich EGF-like domain-containing brain protein | |
CSPG5 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
10675 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SLSTIAEGSHPNVRKLCNTPRTSSPHARALAHYDNVICQDDPSAPHKIQEVLKSCLKEEESFNIQNSMSPKLEGGKGDQADLDVNCL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title