Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurogranin, Rabbit anti-Rat, Human, Polyclonal, Millipore Sigma™

Catalog No. 07425I25UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07425I25UG 25 μg
07425I100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07425I25UG Supplier MilliporeSigma Supplier No. 07425I25UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Neurogranin Antibody, Cat. No. 07-425-I, is a highly specific rabbit polyclonal antibody that targets Neurogranin and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Neurogranin (UniProt: Q92686; also known as Ng, RC3) is encoded by the NRGN gene (Gene ID: 4900) in human. Neurogranin is abundantly expressed in cerebral cortex, hippocampus, amygdala, and striatum. In neurons, it is found concentrated at dendritic spines. It acts as a third messenger and serves as a substrate for protein kinase C-mediated molecular cascades during synaptic development and remodeling. It contains an IQ domain (aa 26-347) and a collagen-like domain (aa 48-78). The IQ domain mediates its interaction with calmodulin and phosphatidic acid (PA). Neurogranin is intrinsically unstructured, however, upon binding with calmodulin, the IQ domain adopts a helical conformation. Neurogranin is phosphorylated at serine 36 by protein kinase C and following phosphorylation neurogranin is unable to bind either calmodulin or PA, which can lead to decline in memory-associated functions. Higher levels of neurogranin have also been reported in brains of elderly and is considered a factor in memory decline, independent of Alzheimer s disease. (Ref.: Javier Dı´ez-Guerra, F. (2010). Life. 62(8): 597-606; Casaletto, KB et al. (2017). Neurology 89(17); 1782-1788).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurogranin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Neurogranin in human and rat brain tissue sections.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Ng;RC3
Gene Symbols NRGN
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 12 amino acids from the C-terminal region of human Neurogranin.
Molecular Weight of Antigen ∽15 kDa observed; 7.62 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001119653
Test Specificity This rabbit polyclonal antibody sepecifically detects Neurogranin. It targets an epitope with in 12 amino acids from the C-terminal region.
Target Species Rat, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.