Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurokinin A Receptor Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980083
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0083 Supplier Bioss Supplier No. BS0123R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene belongs to a family of genes that function as receptors for tachykinins. Receptor affinities are specified by variations in the 5′-end of the sequence. The receptors belonging to this family are characterized by interactions with G proteins and 7 hydrophobic transmembrane regions. This gene encodes the receptor for the tachykinin neuropeptide substance K, also referred to as neurokinin A.

Specifications

Antigen Neurokinin A Receptor
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TACR2
Gene Accession No. P21452
Gene Alias 7 transmembrane receptor; neurokinin 2 receptor; Neurokinin A receptor; NK2; NK-2 receptor; NK2R; NK-2R; NKNAR; seven transmembrane helix receptor; SK receptor; SKR; substance K receptor; Substance-K receptor; Tac2r; Tachykinin 2 (substance K) receptor; tachykinin NK-2 receptor; Tachykinin receptor 2; TACR2
Gene Symbols TACR2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NK2R.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6865
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.