Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurokinin B Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300201G
Description
Neurokinin B Polyclonal specifically detects Neurokinin B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Neurokinin B | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
gamma tachykinin 3, neurokinin beta, neurokinin B-like, neuromedin K, preprotachykinin B, PRO1155, tachykinin 3, tachykinin-3, ZNEUROK1NKNBNKB | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TAC3 | |
A synthetic peptide made to an internal sequence of mouse proNeurokinin B (∼93% homology to the rat sequence). [UniProt# P55099] | |
0.1 mL | |
GPCR, Neuroscience, Neurotransmission | |
6866 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction