Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neuroligin 1 Monoclonal Antibody (N97A/31), FITC
GREENER_CHOICE

Catalog No. PIMA545686
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545686 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545686 Supplier Invitrogen™ Supplier No. MA545686
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse: 99% identity (125/126 amino acids identical). Human: 99% identity (125/126 amino acids identical) >40% identity with Neuroligin-2 and -3. 2 μg/mL of MA5-45686 was sufficient for detection of Neuroligin-1 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 120kDa. Does not cross-react with other Neuroligins. This antibody was formerly sold as clone S97A-31.

This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuroligin 1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N97A/31
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene Nlgn1
Gene Accession No. Q62765, Q8N2Q7, Q99K10
Gene Alias 6330415N05Rik; BB179718; Kiaa1070; MGC45115; mKIAA1070; neuroligin 1; neuroligin I; Neuroligin-1; NL1; NLG 1; Nlg1; NLGN1
Gene Symbols Nlgn1
Host Species Mouse
Immunogen Fusion protein amino acids 718-843 (cytoplasmic C-terminus) of rat Neuroligin-1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116647, 192167, 22871
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.